Learn More
Agilent Technologies LIGHT BLOCK PADS-96WELL PLATES
Catalog No. 50125740
Encompass
Mx4000 Light Blocking Pads for 96-Well Plates is a light blocking device designed for use with 96 well plates to minimize light scattering(signal noise) from one fiber optic to another during optical measurements in the Mx4000 instrument.
Catalog No. 50-125-740 Supplier Agilent Technologies Supplier No. 401266
Mx4000 Light Blocking Pads for 96-Well Plates is a light blocking device designed for use with 96 well plates to minimize light scattering(signal noise) from one fiber optic to another during optical measurements in the Mx4000 instrument.