Learn More
Cenmed™ Light Blue Disposable Scrub Bottom with Elastic Waist
SDP
Catalog No. 21520218
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-520-218 X-Large
21-520-215 Small
21-520-216 Medium
21-520-217 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-520-218 Supplier Cenmed™ Supplier No. C3007018100
  • $639.50
    $404.65 / Case of 100
    Save $234.85
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Single use disposable scrub elastic waist pants provides superior protection against contaminants and prevents bodily fluids from making contact to skin.
  • Optimal coverage, comfort and breathability
  • Ease of range of movement
  • Hypoallergenic
  • SMS Non Woven Material
  • Anti-Static
  • Latex Free
  • 45 g
Specifications
Size X-Large
Gender Unisex
Material SMS
Color Light Blue
Ankle Style Boot Cut
Seam Type Taped
No. of Pockets None
Disposable Single-use
Latex Free Yes
Sterility Non-sterile