Learn More
International Enviroguard™ Light Blue SMS Lab Coat with 3 Pockets, Knit Wrists
Catalog No. 19472821 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-821 2X-Large
19-472-822 3X-Large
19-472-823 4X-Large
19-472-824 Large
19-472-825 Medium
19-472-826 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-472-821 Supplier International Enviroguard™ Supplier No. K2248B2XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
SMS lab coat protects against dust, dirt, grime, and dry particulates in nonhazardous settings.
Light blue SMS lab coat protects against dust, dirt, grime, and other dry particulates in non-hazardous environments. Fabric also helps protect against light liquid splashes and sprays. Fabric is breathable to help keep you cool, so you can work comfortably over long hours. With three roomy pockets, you can store eyeglasses, pens, and other small instruments to keep with you on the go.
  • Three-layer SMS is lightweight and breathable to help prevent heat stress
  • Knit wrists and knit collar
  • 1 left chest pocket and 2 hip pockets
  • Possible applications include pharmaceuticals, labs, healthcare, controlled environments, food processing, and more
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color Blue
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Style Below Hip-length
No. of Pockets 3
Description Blue SMS Lab Coat with 3 pockets, knit wrists, and knit collar
Collar Type Knit
Latex Free Yes
Model K2248B-2XL
Seam Type Serged
Autoclavable Not Autoclavable
No. per Pack 30/Cs.