Learn More
International Enviroguard™ Light Blue SMS Lab Coat with 3 Pockets, Knit Wrists
Shop All International Enviroguard Systems ProductsDescription
Light blue SMS lab coat protects against dust, dirt, grime, and other dry particulates in non-hazardous environments. Fabric also helps protect against light liquid splashes and sprays. Fabric is breathable to help keep you cool, so you can work comfortably over long hours. With three roomy pockets, you can store eyeglasses, pens, and other small instruments to keep with you on the go.
- Three-layer SMS is lightweight and breathable to help prevent heat stress
- Knit wrists and knit collar
- 1 left chest pocket and 2 hip pockets
- Possible applications include pharmaceuticals, labs, healthcare, controlled environments, food processing, and more
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | X-Large |
| Gender | Unisex |
| Material | SMS |
| Color | Blue |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Dry Particulates |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.