Learn More
Stoelting™ Light/Dark Box
SDP
Catalog No. 10000355 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
10-000-355 Mouse
1 options
Catalog No. 10-000-355 Supplier Stoelting™ Supplier No. 63101
Add to Cart
Add to Cart
Stoelting™ Light/Dark Box is a characteristic tool used in the assessment of anxiety.
  • Basic measure is the animal's preference for dark, enclosed places over bright, exposed places.
  • Time spent in the lit (light) half of the arena, and the related exploratory behaviors, are reliable parameters for assessing anxiolytic effects that may be useful in identifying and/or screening of anxiolytic and anxiogenic agents.
Specifications
For Use With (Application) Neuroscience
Animal Mouse