Learn More
Grainger Dayton Air Circulator, 18 in. Blade, 5,700 Cfm, 120 V, 1 ea
Catalog No. 502772323
Encompass_Preferred
This industrial-grade wall-mounted air circulator provides efficient air movement for demanding environments. Designed for stationary use, this oscillating fan features a robust steel housing and guard, paired with durable aluminum blades for reliable performance.
  • 18 in. diameter aluminum blades
  • Two-speed, 1/5 hp permanent split capacitor motor
  • Oscillating design for wide coverage
  • Totally enclosed air over motor enclosure
  • Automatic thermal protection
  • Gloss powder coated steel wire guard
  • 9 ft. power cord with 3 conductors
  • Maximum ambient temperature of 104 °F
Catalog No. 50-277-2323 Supplier Grainger Supplier No. 13V401
This industrial-grade wall-mounted air circulator provides efficient air movement for demanding environments. Designed for stationary use, this oscillating fan features a robust steel housing and guard, paired with durable aluminum blades for reliable performance.
  • 18 in. diameter aluminum blades
  • Two-speed, 1/5 hp permanent split capacitor motor
  • Oscillating design for wide coverage
  • Totally enclosed air over motor enclosure
  • Automatic thermal protection
  • Gloss powder coated steel wire guard
  • 9 ft. power cord with 3 conductors
  • Maximum ambient temperature of 104 °F