Learn More
Ace Glass, Inc. Air motor, light duty, 10, 000rpm max, 1/3hP, 4 in-lbs max torque, 13cfm air consumption, 30-100psi input
SDP
Catalog No. 50977955
Encompass
Air motor, light duty, 10, 000rpm max, 1/3hP, 4 in-lbs max torque, 13cfm air consumption, 30-100psi input
Catalog No. 50-977-955 Supplier Ace Glass, Inc. Supplier No. 1336505
Air motor, light duty, 10, 000rpm max, 1/3hP, 4 in-lbs max torque, 13cfm air consumption, 30-100psi input