Learn More
Bioworld Light Green SF, 25 g
Catalog No. 50199222
Encompass_Preferred
A cytological counterstain used for general plant histology. High Purity Grade EM (630nm, water) >63,500
Catalog No. 50-199-222 Supplier Bioworld Supplier No. 217700241
A cytological counterstain used for general plant histology. High Purity Grade EM (630nm, water) >63,500