Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light Green SF, Yellowish (Certified Biological Stain), Fisher Chemical

Catalog No. O338225 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O3382-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O3382-25

missing translation for 'validMainframeMsg'

Certified for use in histology and cytology

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5141-20-8
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012121
InChI Key DGOBMKYRQHEFGQ-UHFFFAOYSA-L
Synonym Acid Green 5
PubChem CID 21223
ChEBI CHEBI:87065
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 288°C
Bio. Stain Comm. Certification Pass Test
Dye Content Report %
Formula Weight 792.85
Grade Certified Biological Stain
Packaging Glass Bottle
Color Index No. 42095
Color Brown/Red
Quantity 25 g
Physical Form Solid
Chemical Name or Material Light Green SF, Yellowish
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.