Learn More
Abcam Light green SF yellowish, Cytological counterstain, 25G
Catalog No. 502704774
Encompass_Preferred
MW 792.86 g/mol. Triarylmethane dye. Cytological counterstain that can be used as a substitute for aniline blue in Gomori′s one-step trichrome procedure. An important contrast stain component of Papanicolaou (PAP) stains.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4774 Supplier Abcam Supplier No. AB14626825G
MW 792.86 g/mol. Triarylmethane dye. Cytological counterstain that can be used as a substitute for aniline blue in Gomori′s one-step trichrome procedure. An important contrast stain component of Papanicolaou (PAP) stains.
The product is subject to the following: Abcam Restricted Use Statement