Learn More
Electron Microscopy Sciences Light Green SF Yellowish 0.2% in 0.2% Acetic Acid 1 LT
SDP
Catalog No. 5026933
Encompass_Preferred
It is available as 0.2% in 0.2% acetic acid for use in the GM Method, as a fungal stain.
Catalog No. 50-269-33 Supplier Electron Microscopy Sciences Supplier No. 2608506
It is available as 0.2% in 0.2% acetic acid for use in the GM Method, as a fungal stain.