Learn More
Electron Microscopy Sciences Light Green SF Yellowish 0.2% in 0.2% Acetic Acid 500 ML
SDP
Catalog No. 5026934
Encompass_Preferred
It is is available as 0.2% in 0.2% acetic acid for use in the GM Method, as a fungal stain.
Catalog No. 50-269-34 Supplier Electron Microscopy Sciences Supplier No. 2608505
It is is available as 0.2% in 0.2% acetic acid for use in the GM Method, as a fungal stain.