Learn More
Electron Microscopy Sciences Light Green Stock 0.2% 500 ML
SDP
Catalog No. 5030099
Encompass_Preferred
Used in Goldner's Trichrome Method for connective tissue stains – Elastic tissue stain. Stain with a poly-acid step. See Luna, Lee G. Histopathologic Methods and Color Atlas of Special Stains and Tissue Artifacts. Johnson Printers, Downerts Grove, IL p151-152, c 1992.
Catalog No. 50-300-99 Supplier Electron Microscopy Sciences Supplier No. 2638607
Used in Goldner's Trichrome Method for connective tissue stains – Elastic tissue stain. Stain with a poly-acid step. See Luna, Lee G. Histopathologic Methods and Color Atlas of Special Stains and Tissue Artifacts. Johnson Printers, Downerts Grove, IL p151-152, c 1992.