Learn More
WELCH ALLYN INC 78900 Series KleenSpec Corded Illumination System
Catalog No. NC1591995
Encompass
The 78900 Series KleenSpec Corded Illumination System, with halogen technology, provides bright, long-lasting light to the exam or procedure area. Ideal for facilities that require an illumination system that requires high-level disinfection.
Catalog No. NC1591995 Supplier WELCH ALLYN INC Supplier No. 78900
The 78900 Series KleenSpec Corded Illumination System, with halogen technology, provides bright, long-lasting light to the exam or procedure area. Ideal for facilities that require an illumination system that requires high-level disinfection.