Learn More
ACROBiosystems ACROBiosystems Human LIGHT / TNFSF14 Protein, His Tag (active trimer) (MALS verified)
Catalog No. 501956402
Encompass_Preferred
Recombinant Protein;100UG;Human LIGHT, His Tag (active trimer) (MALS verified) (LIT-H5242) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 58.3 kDa. The protein migrates as 55-65 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 5 μg/mL (100 μL/well) can bind Human LIGHT, His Tag (active trimer) (MALS verified) (Cat. No. LIT-H5242) with a linear range of 16-63 ng/mL (QC tested).;Immobilized Human DcR3, Fc Tag (Cat. No. TNB-H5255) at 5 μg/mL (100 μL/well) can bind Human LIGHT, His Tag (active trimer) (MALS verified) (Cat. No. LIT-H5242) with a linear range of 8-63 ng/mL (Routinely tested).
Catalog No. 50-195-6402 Supplier ACROBiosystems Supplier No. LITH5242100UG
Recombinant Protein;100UG;Human LIGHT, His Tag (active trimer) (MALS verified) (LIT-H5242) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1) trimer Design.;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 58.3 kDa. The protein migrates as 55-65 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 5 μg/mL (100 μL/well) can bind Human LIGHT, His Tag (active trimer) (MALS verified) (Cat. No. LIT-H5242) with a linear range of 16-63 ng/mL (QC tested).;Immobilized Human DcR3, Fc Tag (Cat. No. TNB-H5255) at 5 μg/mL (100 μL/well) can bind Human LIGHT, His Tag (active trimer) (MALS verified) (Cat. No. LIT-H5242) with a linear range of 8-63 ng/mL (Routinely tested).