Learn More
PCE AMERICAS INC Lux Meter measures from 1000 to 400,000 Lux with a resolution of 0.1 lux.
SDP
Catalog No. NC3654895
Encompass_Preferred
Illuminometer with external probe / Sampling rate 4 Hz / Measuring range 0 ... 400 kLux / Micro-USB interface / Units: lux, footcandle. The illuminometer has additional functions so that an analysis of the illuminance can be carried out. With the additional functions of the illuminometer the largest and smallest measured value can be output using the "MIN" and "MAX" buttons. In addition, the displayed measured value can be set in relation to a reference using the "REL" button. With the "PEAK" function, the illuminometer records peak values. The illuminometer has a sampling rate of 10 µs. The PCE-LMD 10 also has a data memory of 32,000 measured values. The measured values ​​from the illuminometer can be read out and analyzed on the PC after a measurement. A live measurement on the PC with the illuminometer is also possible. The data from the illuminometer can be exported in CSV format for further processing.
Catalog No. NC3654895 Supplier PCE AMERICAS INC Supplier No. PCELMD 5
Illuminometer with external probe / Sampling rate 4 Hz / Measuring range 0 ... 400 kLux / Micro-USB interface / Units: lux, footcandle. The illuminometer has additional functions so that an analysis of the illuminance can be carried out. With the additional functions of the illuminometer the largest and smallest measured value can be output using the "MIN" and "MAX" buttons. In addition, the displayed measured value can be set in relation to a reference using the "REL" button. With the "PEAK" function, the illuminometer records peak values. The illuminometer has a sampling rate of 10 µs. The PCE-LMD 10 also has a data memory of 32,000 measured values. The measured values ​​from the illuminometer can be read out and analyzed on the PC after a measurement. A live measurement on the PC with the illuminometer is also possible. The data from the illuminometer can be exported in CSV format for further processing.