Learn More
Cole-Parmer Extech EA30 Easyview Wide-Range Light Meter
Catalog No. 50992894 Shop All Cole-Parmer Products
Encompass
This meter measures light intensity up to 400,000 lux in five ranges and 40,000 footcandles in four ranges. Unit is color and cosine corrected. Large LCD simultaneously displays readings numerically and in a bar graph format. Meter utilizes a precision silicon photodiode sensor to obtain readings.
Catalog No. 50-992-894 Supplier Cole-Parmer Supplier No. 0158865
This meter measures light intensity up to 400,000 lux in five ranges and 40,000 footcandles in four ranges. Unit is color and cosine corrected. Large LCD simultaneously displays readings numerically and in a bar graph format. Meter utilizes a precision silicon photodiode sensor to obtain readings.