Learn More
Electron Microscopy Sciences Light Microscopy Perfect Loop Only
SDP
Supplier:  Electron Microscopy Sciences 70942
Encompass_Preferred
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases. For light microscopy (large sections). Outside diameter of loop is 7mm.
Catalog No. 50-930-556