Learn More
Aqua Solutions Mineral Oil Light Laboratory Grade (80-90 SUS @100°F) (500ML)
SDP
Catalog No. NC9849651
Encompass_Preferred
Mineral Oil Light Laboratory Grade (80-90 SUS @100°F) (500ML)
Catalog No. NC9849651 Supplier Aqua Solutions Supplier No. M7113500ML
Mineral Oil Light Laboratory Grade (80-90 SUS @100°F) (500ML)