Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Light module for Plant Growth

Catalog No. 15500207 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
Plant growth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
15-500-207 Plant growth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 15-500-207 Supplier Thermo Scientific™ Supplier No. 50158128

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Material: Stainless Steel; For Use With: All Thermo Scientific and Forma vertical light chambers - models 3906, 3909, 3928, 3942, 3943

Light module with 7 fluorescent bulbs. Choose module based on your application: ICH photostability testing, plant growth, animal hatching. Fluorescent bulbs vary based on chosen module to address specific light requirement of application. Please replace a

Light module for sample exposure with defined light. Please choose from three application-based versions:

 

  • For ICH stability testing, light of module allows photo stability test according to ICH guidelines in 170 hours. Replacement bulb kit 50158286.
  • For plant growth, light of module is high in blue and red for ideal growth of plants. Replacement bulb kit 50158287.
  • For animal hatching, light of module mimics daylight. Replacement bulb kit 50158287.
    NOTE: Please exchange all 7 bulbs at once for even light distribution.
TRUSTED_SUSTAINABILITY

Specifications

Description Light module plant growth - for use with vertical light chambers
For Use With (Equipment) All Thermo Scientific and Forma vertical light chambers - models 3906, 3909, 3928, 3942, 3943
Lighting LED
Material Stainless Steel
Type Light Module
For Use With (Application) Plant growth
Frame Material Stainless steel
Includes 7 fluorescent bulbs
Warranty 2 years
Shipping Weight (English) 59.5 lbs
Shipping Weight (Metric) 27 kg
Weight (English) 27.8 lbs
Weight (Metric) 12.6 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.