Learn More
Roboz Surgical LIGHT OP SCS 5IN STR S/S
SDP
Catalog No. NC9341918
Encompass
Light Operating Scissors; Straight; Sharp-Sharp; 40mm Blade Length; 5 in. Overall Length
Catalog No. NC9341918 Supplier Roboz Surgical Supplier No. RS6752
Light Operating Scissors; Straight; Sharp-Sharp; 40mm Blade Length; 5 in. Overall Length