Learn More
Electron Microscopy Sciences Light Pipe Scintillator JEOL JSM-35 Quartz 0.5" Collar, 60mm
SDP
Catalog No. 5026942
Encompass
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT. We coat the scintillator material directly on the light pipe which improves the signal. Price includes one Re-coating Certificate. Re-coating is typically needed twice per year. High voltage wire and corona ring are included.
OAL: 60mm
Catalog No. 50-269-42 Supplier Electron Microscopy Sciences Supplier No. 8200560
Add to Cart
Add to Cart
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT. We coat the scintillator material directly on the light pipe which improves the signal. Price includes one Re-coating Certificate. Re-coating is typically needed twice per year. High voltage wire and corona ring are included.
OAL: 60mm