Learn More
Electron Microscopy Sciences Light Pipe Scintillator Cambridge Quartz, Up To 200 mm
SDP
Catalog No. 5026950
Encompass
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT. We coat the scintillator material directly on the light pipe which improves the signal. Price includes one Re-coating Certificate. Re-coating is typically needed twice per year. Substrate: optical grade quartz, straight, high voltage wire and corona ring are included.
OAL: Up To 200 mm
Catalog No. 50-269-50 Supplier Electron Microscopy Sciences Supplier No. 82000X
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT. We coat the scintillator material directly on the light pipe which improves the signal. Price includes one Re-coating Certificate. Re-coating is typically needed twice per year. Substrate: optical grade quartz, straight, high voltage wire and corona ring are included.
OAL: Up To 200 mm