Learn More
Anton Paar Light-scattering instrument for characterizing particlesin liquid dispersions. Measures particle size, zeta potential,electrophoretic mobility, molecular mass and transmittance
Catalog No. 5000580464
Encompass_Preferred
Litesizer DLS 301 specificationsMeasurement principle: Dynamic Light Scattering (DLS)Measuring range: 0.3 nm to 15 μm* (particle diameter)Measurement angles: 90°Min. concentration: 1 mg/mL (lysozyme)Max. concentration: 40 % w/v (sample-dependent)Min. sample volume: 1.5 µLAccuracy: Better than +/-2 % on NIST traceable standardsRepeatability: Better than +/-2 % on NIST traceable standardsGeneral specificationsLight source: Semiconductor laser diode / 40 mW, 658 nmLaser warm-up time: 6 minTemperature control range: 0 °C to 120 °CAmbient operating temperature: 10 °C to 35 °CHumidity: Up to 80 % non-condensingDimensions: (W, D, H)450 mm, 505 mm, 135 mmWeight:Approx. 18 kg (40 lb)
Catalog No. 50-005-80464 Supplier Anton Paar Supplier No. 394443
Please contact your sales representative for a quote or further ordering instructions.
Litesizer DLS 301 specificationsMeasurement principle: Dynamic Light Scattering (DLS)Measuring range: 0.3 nm to 15 μm* (particle diameter)Measurement angles: 90°Min. concentration: 1 mg/mL (lysozyme)Max. concentration: 40 % w/v (sample-dependent)Min. sample volume: 1.5 µLAccuracy: Better than +/-2 % on NIST traceable standardsRepeatability: Better than +/-2 % on NIST traceable standardsGeneral specificationsLight source: Semiconductor laser diode / 40 mW, 658 nmLaser warm-up time: 6 minTemperature control range: 0 °C to 120 °CAmbient operating temperature: 10 °C to 35 °CHumidity: Up to 80 % non-condensingDimensions: (W, D, H)450 mm, 505 mm, 135 mmWeight:Approx. 18 kg (40 lb)