Learn More
Hach Company Replacement Light Shield, DR3900
Supplier:  Hach Company LZV849
Encompass
The light shield protects the optic from stray light if the round cell compartment is used, e.g with TNTplus chemistry.
Catalog No. 50-199-7947