Learn More
AdipoGen LIGHT, Soluble (human) (rec.) (10 µg)
SDP
Supplier:  AdipoGen AG40B0009C010
Encompass_Preferred
Protein. Human LIGHT (aa 89-240) is fused at the N-terminus to a FLAG-tag. Source: CHO cells. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Binds to human and mouse LTbetaR and human HVEM and DcR3. Purity: >95% (SDS-PAGE). LIGHT (HVEML; CD258) is a cytokine that binds to TNFRSF3/LTBR. It activates NF-kappaB, stimulates the proliferation of T cells, and inhibits growth of the adenocarcinoma HT-29. Acts as a receptor for Herpes simplex virus.
Catalog No. 50-205-3078