Learn More
Enzo Life Sciences LIGHT (soluble) (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522018C010
Encompass_Preferred
Alternative name: HVEML, TNFSF 14, CD258. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human LIGHT (aa 89-240) is fused at the N-terminus to a linker peptide (10 aa) and a FLAG-tag. Specificity: Binds to human and mouse LTβR and human HVEM and DcR3. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9368