Learn More
Caron Products And Services Light tight door cover
Supplier:  Caron Products And Services DOOR312
Encompass_Preferred
Light tight door cover for 934 L (33 ft3) series chambers/incubators (with glass). Factory installed at the time of order or can be retro-fitted in the field by a qualified technician.
Catalog No. NC2589292
Add to Cart
Request a Quote