Learn More
Valutek™ Light Weight Polyester Wiper, Cold Knife Cut
Catalog No. 191502020
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
Catalog No. Dimensions (L x W)
19-150-2020 9 x 9 in.
1 options
This item is not returnable. View return policy
Catalog No. 19-150-2020 Supplier Valutek™ Supplier No. VTPNWLW99
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed from light-weight, 100% continuous filament polyester in a double-knit, no-run, interlock pattern with cold knife cut edges. This wiper is a clean and moderately absorbent wiper with low levels of particulate and extractable counts.
  • 100% continuous filament, double-knit polyester fiber
  • Cold knife cut edge
  • Moderate absorbency and abrasion resistance
  • Chemical compatible with IPA and other common solvents
  • Low levels of particulate and extractable counts
  • Light weight material reduces both actual cost and the landfill waste impact
  • Laundered and packaged in a cleanroom, all Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 100 -1000 (ISO 5-6) critical environment
  • It is also commonly used for a wide variety of applications such as spill control, general wiping, breakdown cleaning in fab and suite construction, cleaning medical device products, cleaning and burnishing magnetic media surfaces, and wiping sensitive surfaces.
Specifications
Material Polyester
Dimensions (L x W) 9 x 9 in.
Sterility Non-sterile