Learn More
Roche Diagnostics LIGHTCYCLER 480 PROBES MASTER
Supplier:  Roche Diagnostics 04887301001
Encompass
LightCycler 480 Probes Master - 10 x 5 ml (2x conc.) (approx. 10 x 500 reactionsof 20 uL final reaction volume)
Catalog No. 50-720-3232