Learn More
Roche Diagnostics LightCycler 480 SYBR Green I Master
Supplier:  Roche Diagnostics 04707516001
Encompass
LightCycler 480 SYBR Green I Master - 5 x 1 ml (2x conc.) (approx. 500 reactions of 20uL final reaction volume)
Catalog No. 50-720-3180