Learn More
Roche Diagnostics LIGHTCYCLER 480 XENON BULB
Supplier:  Roche Diagnostics 04686136001
Encompass
The LightCycler 480 Xenon Lamp is a replacement part for the LightCycler 480 Instrument. The lamp can be easily replaced by the user within a few minutes.
Catalog No. 50-587-965