Learn More
Percival Scientific Percival I-36DL Dew Formation Chamber, Programmable SciWhite LED Lighting, Precision Temperature Control, 1 Unit
SDP
Catalog No. NC3253384
Encompass_Preferred
The Percival Scientific I-36DL Lighted Dew Formation Chamber is designed for precise environmental control in plant pathology studies. It simulates natural dew formation with accurate temperature regulation, featuring a robust stainless steel and aluminum build, corrosion resistance, and programmable SciWhite LED lighting. Controlled by the IntellusUltra system, it ensures stable conditions for repeatable research outcomes, making it a valuable tool for experimental plant inoculation processes.
Catalog No. NC3253384 Supplier Percival Scientific Supplier No. I36DL
Add to Cart
Add to Cart
Request a Quote
The Percival Scientific I-36DL Lighted Dew Formation Chamber is designed for precise environmental control in plant pathology studies. It simulates natural dew formation with accurate temperature regulation, featuring a robust stainless steel and aluminum build, corrosion resistance, and programmable SciWhite LED lighting. Controlled by the IntellusUltra system, it ensures stable conditions for repeatable research outcomes, making it a valuable tool for experimental plant inoculation processes.