Learn More
Electron Microscopy Sciences Lighted Forceps with 20 cm Serrated Blunt Tips
SDP
Catalog No. 501902131
Encompass
See your samples and your embeddings in a whole new light! These lighted forceps ease identification and location of otherwise undetectable tissue fragments, while reducing embedding time. Increases the visibility of specimen allowing for more accurate work. Made of German stainless steel, are submersible and light longevity approx. 20,000 hours.
Size: 20 cm
Tips: Serrated Blunt Tip
Catalog No. 50-190-2131 Supplier Electron Microscopy Sciences Supplier No. 7790506
See your samples and your embeddings in a whole new light! These lighted forceps ease identification and location of otherwise undetectable tissue fragments, while reducing embedding time. Increases the visibility of specimen allowing for more accurate work. Made of German stainless steel, are submersible and light longevity approx. 20,000 hours.
Size: 20 cm
Tips: Serrated Blunt Tip