Learn More
Grainger Lutron Skylark Dimmer, 3-Way, 3-Wire Fluorescent/LED, 277 V, Ivory, 1 ea
Catalog No. 502795131
Encompass_Preferred
The Skylark® series 3-way designer-style dimmer is designed for use with 3-wire fluorescent ballasts or LED drivers. This unit supports a voltage rating of 277 V AC and an amperage rating of 6 A, providing reliable lighting control in commercial and residential settings.
  • Designed for 3-wire fluorescent or LED applications
  • Preset start type with slider control for convenient dimming adjustment
  • Operates at 277 V AC with a 6 A load capacity
  • Temperature rated for 0°C to 40°C environments
  • Features a durable plastic construction in an ivory finish
  • Designed for vertical wall mounting
  • Compatible with copper wire only
Catalog No. 50-279-5131 Supplier Grainger Supplier No. 19YP97
The Skylark® series 3-way designer-style dimmer is designed for use with 3-wire fluorescent ballasts or LED drivers. This unit supports a voltage rating of 277 V AC and an amperage rating of 6 A, providing reliable lighting control in commercial and residential settings.
  • Designed for 3-wire fluorescent or LED applications
  • Preset start type with slider control for convenient dimming adjustment
  • Operates at 277 V AC with a 6 A load capacity
  • Temperature rated for 0°C to 40°C environments
  • Features a durable plastic construction in an ivory finish
  • Designed for vertical wall mounting
  • Compatible with copper wire only