Learn More
Grainger Lutron Lighting Dimmer, 1-Pole and 3-Way, 120V AC, Ivory
Catalog No. 502795923
Encompass_Preferred
This lighting dimmer offers advanced control for dimmable LED, CFL, halogen, and incandescent bulbs. Designed for single-pole or 3-way applications, this device features a linear slider for precise light level adjustment and a rocker switch to turn lights on and off. The unit includes power-failure memory, returning lights to the previous level, and a mechanical air-gap switch to disconnect power for bulb replacement.
  • Provides 150 W capacity for CFL and LED bulbs and 600 W for incandescent or halogen bulbs
  • Compatible with 120 V AC, 60 Hz electrical systems
  • Features a low-end adjustment dial to accommodate a wide range of bulb types
  • Constructed from resin with a gloss finish in ivory
  • UL Listed to US and Canadian safety requirements (UL 1472 and CSA C22.2 184.1) and NOM certified
  • No neutral wire required for installation
Catalog No. 50-279-5923 Supplier Grainger Supplier No. 19YP99
This lighting dimmer offers advanced control for dimmable LED, CFL, halogen, and incandescent bulbs. Designed for single-pole or 3-way applications, this device features a linear slider for precise light level adjustment and a rocker switch to turn lights on and off. The unit includes power-failure memory, returning lights to the previous level, and a mechanical air-gap switch to disconnect power for bulb replacement.
  • Provides 150 W capacity for CFL and LED bulbs and 600 W for incandescent or halogen bulbs
  • Compatible with 120 V AC, 60 Hz electrical systems
  • Features a low-end adjustment dial to accommodate a wide range of bulb types
  • Constructed from resin with a gloss finish in ivory
  • UL Listed to US and Canadian safety requirements (UL 1472 and CSA C22.2 184.1) and NOM certified
  • No neutral wire required for installation