Learn More
Grainger Lutron Skylark C·L Dimmer, 1-Pole/3-Way, Gloss White
Catalog No. 502796287
Encompass_Preferred
The Skylark LED+ dimmer is designed for the superior control of dimmable LED, CFL, halogen, and incandescent lighting. Featuring a convenient rocker switch with a linear-slide dimmer, this unit maintains preferred lighting levels and includes power-failure memory to return to the last setting upon restoration. It is engineered for single-pole or 3-way applications and rated for 120 V at 60 Hz operation.
  • Controls up to 150 W of dimmable LED or CFL bulbs or up to 600 W of incandescent or halogen bulbs.
  • Compatible with up to 6 forward-phase LED drivers.
  • Includes a mechanical air-gap switch to disconnect load power.
  • Features low-end adjustment to accommodate various bulb types.
  • 100% factory tested.
  • UL listed to US and Canadian safety requirements (UL 1472 / CSA C22.2 184.1).
  • Suitable for indoor use with an operating temperature range of 0°C to 40°C.
Catalog No. 50-279-6287 Supplier Grainger Supplier No. 19YP98
The Skylark LED+ dimmer is designed for the superior control of dimmable LED, CFL, halogen, and incandescent lighting. Featuring a convenient rocker switch with a linear-slide dimmer, this unit maintains preferred lighting levels and includes power-failure memory to return to the last setting upon restoration. It is engineered for single-pole or 3-way applications and rated for 120 V at 60 Hz operation.
  • Controls up to 150 W of dimmable LED or CFL bulbs or up to 600 W of incandescent or halogen bulbs.
  • Compatible with up to 6 forward-phase LED drivers.
  • Includes a mechanical air-gap switch to disconnect load power.
  • Features low-end adjustment to accommodate various bulb types.
  • 100% factory tested.
  • UL listed to US and Canadian safety requirements (UL 1472 / CSA C22.2 184.1).
  • Suitable for indoor use with an operating temperature range of 0°C to 40°C.