Learn More
ACROBiosystems Recombinant Protein;Human LIGHT / TNFSF14 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;1MG;LIT,CD258 antigen,CD258,HVEM-L,LIGHT,LTg,TNFSF14,TR2,HVEML,HVEM,TNFRSF14
Catalog No. 501657742
Encompass_Preferred
Recombinant Protein;Human LIGHT / TNFSF14 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (LIT-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-165-7742 Supplier ACROBiosystems Supplier No. LITH52561MG
Recombinant Protein;Human LIGHT / TNFSF14 Protein, Mouse IgG2a Fc Tag, low endotoxin;1MG;Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (LIT-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) with a linear range of 0.3-5 ng/mL (QC tested).