Learn More
Cole-Parmer Lightnin 829448 316 SS four-blade impeller ( A-200), 2.0" dia x 5/16" bore dia
Catalog No. NC1655267 Shop All Cole-Parmer Products
Encompass
Four-blade impellers are turbine impellers that have a 45 degree pitch and produce a semi-axial flow
Catalog No. NC1655267 Supplier Cole-Parmer Supplier No. 0456160
Four-blade impellers are turbine impellers that have a 45 degree pitch and produce a semi-axial flow