Learn More
Cole-Parmer Lightnin 146835316 316 SS four-blade impeller ( A-200), 3.0" dia x 5/16" bore dia
Catalog No. NC0495073 Shop All Cole-Parmer Products
Encompass
Four-blade impellers are turbine impellers that have a 45 degree pitch and produce a semi-axial flow
Catalog No. NC0495073 Supplier Cole-Parmer Supplier No. 0456060
Four-blade impellers are turbine impellers that have a 45 degree pitch and produce a semi-axial flow