Learn More
Cole-Parmer Lightnin 809411 High-Efficiency Axial Flow Impeller, 5/16" Bore; 3.8" Dia
Catalog No. NC1753787 Shop All Cole-Parmer Products
Encompass_Preferred
High-efficiency axial flow impellers have laser designed paddles to develop 50% more mixing action for high-flow, low-shear mixing.
Catalog No. NC1753787 Supplier Cole-Parmer Supplier No. 0456022
High-efficiency axial flow impellers have laser designed paddles to develop 50% more mixing action for high-flow, low-shear mixing.