Learn More
Cole-Parmer Lightnin Labmaster GP2 Digital Laboratory Mixer Kit, Universal Mount; 115/230 VAC, 50/60Hz
Catalog No. NC1617930 Shop All Cole-Parmer Products
Encompass
Designed for multifunctional open tank applications, these Lightnin GP2 mixers have speeds up to 2000 rpm and 144 in-oz maximum torque. Digital control and feedback system coupled with Lighnin impellers produce consistent results that is critical for validation and scale-up operations. Through-shaft design accepts 3/8" (10 mm) dia shaft and offers quick shaft length adjustments, resulting in less downtime. Operate mixers in clockwise or counterclockwise direction or in agitate cycle along with a timer that stops the mixer when time is complete. Mixers feature brushless DC motor, automatic overload protection, and overtemperature protection to ensure low maintenance and long service life. Chemical-resistant, IP42-rated, keypad makes cleanup simple.
Catalog No. NC1617930 Supplier Cole-Parmer Supplier No. 5080012
Designed for multifunctional open tank applications, these Lightnin GP2 mixers have speeds up to 2000 rpm and 144 in-oz maximum torque. Digital control and feedback system coupled with Lighnin impellers produce consistent results that is critical for validation and scale-up operations. Through-shaft design accepts 3/8" (10 mm) dia shaft and offers quick shaft length adjustments, resulting in less downtime. Operate mixers in clockwise or counterclockwise direction or in agitate cycle along with a timer that stops the mixer when time is complete. Mixers feature brushless DC motor, automatic overload protection, and overtemperature protection to ensure low maintenance and long service life. Chemical-resistant, IP42-rated, keypad makes cleanup simple.