Learn More
Novus Biologicals™ Lightning-Link (R) Fluorescein Antibody Labeling Kit
SDP
Catalog No. 7070005
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
70-700-05 100 μg
1 options
Catalog No. 70-700-05 Supplier Novus Biologicals™ Supplier No. 7070005
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
Antibody labeling kit
Lightning-Link™ antibody labeling kits enable the direct labeling of antibodies, proteins, peptides or other biomolecules for use in R&D applications, drug discovery and the development of diagnostic kits.
Kit Contents:
  • 1 vial of LL-modifier reagent
  • 1 vial of LL-quencher FD reagent
  • 1 glass vial of Lightning-Link™ mix
Specifications
Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C