Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit
SDP

Catalog No. 7630030
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-300-30 30 μg
76-300-05 100 μg
76-300-10 300 μg
76-300-15 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 76-300-30 Supplier Novus Biologicals™ Supplier No. 7630030

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Applications: Flow Cytometry, Electron Microscopy, Flow (Cell Surface)

PerCP/Cy5.5 is a tandem conjugate. The PerCP is excited at 482nm and functions as an energy donor for the Cy5.5. Energy is transferred from the PerCP to the Cy5.5 via energy resonance transfer. The Cy5.5 emits the energy received from the PerCP in the form of long wavelength light at 700nm.

Order Info

Quantity

Specifications

Product Type PerCP-Cy5.5 Antibody Labeling Kit
Content And Storage Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent. Store at -20°C. Avoid freeze-thaw cycles.
Conjugate PerCP/Cy5.5
Quantity 30 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.