Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto633 Antibody Labeling Kit
SDP
Catalog No. 3530005
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
35-300-05 100 μg
1 options
Catalog No. 35-300-05 Supplier Novus Biologicals™ Supplier No. 3530005
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
Specifications
Quantity 100 μg
Product Type Antibody Labeling Kit
Content And Storage -20°C