Learn More
Sigma Aldrich Fine Chemicals Biosciences LightOx PhotoReact 3651EA
Catalog No. 502908663
Encompass
The LightOx PhotoReact 365 is a benchtop instrument that provides even directed and reproducible UV illumination at 365 nm suitable for photochemical and photobiological reactions. The user can control parameters of intensity and energy delivery rate and time. LED health over time is monitored usingan in-built UV sensor enabling the user to produce rapid reproducible results. This instrument is suitable for high-throughput screening in multiwell plates providing researchers with a tool to screen high numbers of compounds and reactions essential for drug discovery.Features and Benefits.nbspThe PhotoReact 365 is a bench top instrument for the acceleration of photochemical and photobiological reactions.nbspInstrument suitable for use with a standard size culture multi-well plate (85.5 mm x 128 mm) or single 95 mm round dish.nbspUser selected light output from 0 mW/cm2 to maximum (approx. 13.0 mW/cm2).nbspVariance less than 1mW/cm2 (at 100% power) and 0.5 mW/cm2 (at 50% power) across th
Catalog No. 50-290-8663 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7440611EA
Add to Cart
Add to Cart
Request a Quote
The LightOx PhotoReact 365 is a benchtop instrument that provides even directed and reproducible UV illumination at 365 nm suitable for photochemical and photobiological reactions. The user can control parameters of intensity and energy delivery rate and time. LED health over time is monitored usingan in-built UV sensor enabling the user to produce rapid reproducible results. This instrument is suitable for high-throughput screening in multiwell plates providing researchers with a tool to screen high numbers of compounds and reactions essential for drug discovery.Features and Benefits.nbspThe PhotoReact 365 is a bench top instrument for the acceleration of photochemical and photobiological reactions.nbspInstrument suitable for use with a standard size culture multi-well plate (85.5 mm x 128 mm) or single 95 mm round dish.nbspUser selected light output from 0 mW/cm2 to maximum (approx. 13.0 mW/cm2).nbspVariance less than 1mW/cm2 (at 100% power) and 0.5 mW/cm2 (at 50% power) across th