Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lightox(Tm)17

Catalog No. LIGHTOX17 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LIGHTOX17 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LIGHTOX17 Supplier Sigma Aldrich Supplier No. LIGHTOX171EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fluorescence microscopy, fluorescence spectroscopy.Chemical Equivalency 1ea=1mgLightOx is a trademark of LightOx Ltd.

Specifications

Density 0.914 g/mL at 25°C
Molecular Formula C6H10O
Linear Formula CH3C5H7(=O)
Quantity 1 Ea.
MDL Number MFCD00064147
Molecular Weight (g/mol) 98.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.