Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lightox(Tm)26

Catalog No. LIGHTOX26 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LIGHTOX26 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LIGHTOX26 Supplier Sigma Aldrich Supplier No. LIGHTOX261EA
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fluorescence microscopy, fluorescent retinoid binding assay, fluorescence spectroscopy.Strong ligand for retinoic acid receptor proteinsChemical Equivalency 1ea=1mgLightOx is a trademark of LightOx Ltd.

Specifications

CAS 2342579-17-1
Molecular Formula C12H18N2O4S
MDL Number MFCD28053568
Quantity 1 g
Synonym Ethyl N-tert-butyl-4-nitrobenzenesulfonimidate; Maricich ethylation reagent
Molecular Weight (g/mol) 286.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.