Learn More
Description
- Designed for easy handling and cost efficiency, with a reliable seal that protects contents while providing quick access when needed.
- Ideal for low to moderate weight items where flexibility and visibility are important.
- Multiple sizes for different substances
- Resealable
- Made of 2 mm polyethylene
Specifications
Specifications
| Dimensions (L x W) | 13 x 18 in. |
| Product Type | Evidence Bag |
| Color | Transparent |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
