Learn More
National Safety Apparel™ Lightweight UltraSoft FR Balaclava
SDP
Catalog No. 17001707 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Khaki
Catalog No.
17-001-707 Color: Khaki
1 options
Catalog No. 17-001-707 Supplier National Safety Apparel™ Supplier No. H11RX
Designed for maximum comfort and flexibility, it is constructed from 6.5 oz. Khaki UltraSoft fabric with flat seams
  • Clothing Type: Hoods
  • Khaki
  • Compliance: Cat 2
  • Country of Origin: USA - Subject to change
  • 6.5 oz. fabric weight
  • Indura ultra soft material
Specifications
Size Medium
Material Westex™ UltraSoft™ Rib
Color Khaki
Seam Type Stitched
Arc Flash Rating 12 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462